Imprimer

Anti-PI 3 Kinase p85 alpha Rabbit Monoclonal Antibody [clone: EP380Y]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB40755-100 AB40755-1ML AB40755-20UL
ABCAAB40755-100EA 707 EUR
ABCAAB40755-100 ABCAAB40755-1ML ABCAAB40755-20UL
Anti-PI 3 Kinase p85 alpha Rabbit Monoclonal Antibody [clone: EP380Y]
Anticorps
Rabbit monoclonal [EP380Y] to PI 3 Kinase p85 alpha.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase p85 alpha
Clonality: Monoclonal
Clone: EP380Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR