Imprimer

Anticorps monoclonal de lapin anti-Calcitonine [clone : SP17]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB16697-500 AB16697-100 AB16697-1
ABCAAB16697-500EA 913 EUR
ABCAAB16697-500 ABCAAB16697-100 ABCAAB16697-1
Anticorps monoclonal de lapin anti-Calcitonine [clone : SP17]
Anticorps
Rabbit monoclonal [SP17] to Calcitonin.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Calcitonin
Clonality: Monoclonal
Clone: SP17
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR