Imprimer

Anti-6xHis Monoclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA100013EA 484 EUR
ORIGTA100013
Anti-6xHis Monoclonal Antibody
Anticorps
Type: Tag
Antigen: 6XHis
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR