Imprimer

Anti-FCGR2/3 Rat monoclonal antibody [clone: 2.4G2]

Fournisseur: Tonbo Biosciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

40-0161-U500 40-0161-U100 40-0161-M001
40-0161-U500EA 202 EUR
40-0161-U500 40-0161-U100 40-0161-M001
Anti-FCGR2/3 Rat monoclonal antibody [clone: 2.4G2]
Anticorps
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). As these are receptors for the Fc portion of mouse IgG, they may also bind laboratory antibody preparations and products used in a variety of cell analysis protocols such as flow cytometry, immunohistochemistry and functional cell assays. The 2.4G2 antibody is therefore widely used as a pre-treatment reagent to block binding of specific antibodies of interest, e.g. fluorescently conjugated antibodies, to Fc receptors via their Fc domains and contributing to “non-specific” staining.

Type: Primary
Antigen: FCGR2/3 (Fc fragment of IgG, low affinity II/III)
Clonality: monoclonal
Clone: 2.4G2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR