Imprimer

Anti-CD44 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: IM7]

Fournisseur: Tonbo Biosciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

35-0441-U025 35-0441-U100 35-0441-U500
35-0441-U025EA 27.7 EUR
35-0441-U025 35-0441-U100 35-0441-U500
Anti-CD44 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: IM7]
Anticorps
The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or “standard” form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.

The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone: IM7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR