Imprimer
Container, Securitainer®
Fournisseur: COVERA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
49X75/01 C49/01 49X58/01 26X51/01 49X85/01 62X90/01 C26/01 C62/01 35X82/01 35X52/01 C109/01 62X113/01 109X100/01
COVE49X75/01EA 233 EUR
COVE49X75/01 COVEC49/01 COVE49X58/01 COVE26X51/01 COVE49X85/01 COVE62X90/01 COVEC26/01 COVEC62/01 COVE35X82/01 COVE35X52/01 COVEC109/01 COVE62X113/01 COVE109X100/01
Container, Securitainer®
Récipients de collecte d'échantillon Conteneurs à usage universel
SECURITAINER®, made of white PP is an effective security container. It has two main uses: as an individual transport container for glass vials and as packaging for materials and substances sensitive to dampness.
  • The tamper-proof seal guarantees that the packaging has not been opened-providing increased product security
  • The seal prevents manipulation of vials, labels and their contents
  • The strong, light construction protects the contents effectively
  • Easy to handle
  • Lids to be ordered seperately