Imprimer
Combined pH electrodes, InPro® 4260
Fournisseur: Mettler - Toledo
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
52002988 52002986 52002987 52002989
METN52002988EA 0 EUR
METN52002988 METN52002986 METN52002987 METN52002989
Combined pH electrodes, InPro® 4260
Électrodes Électrodes pH
These combination pH electrodes with built-in temperature sensor are designed for highly demanding chemical processes.
  • Especially adapted for low ionic content media and organic solvents
  • Insensitive to flow variations
  • Low maintenance pH electrode-no refilling of electrolyte
  • Integrated temperature sensor now positioned inside the glass membrane part offers increased performance
  • Open junction between sample medium and reference electrolyte (no diaphragm – no clogging) allows reliable measurements in heavily contaminated process media, in suspensions and emulsions, and in solutions containing protein
The electrodes offer enhanced performance and a wider application field, especially in harsh chemical processes, due to a new composition of the polymer reference system as a successor to the XEROLYT® polymer.
Reference system: XEROLYT® EXTRA
Certifications: Ex certification: Ex ia IIC T6/T5/T4/T3 Ga/Gb, SEV 14 ATEX 0168 X, IECEx SEV 14.0025X
FM certification: IS /I, II, III /1/ ABCDEFG/ T6