Imprimer
Cell culture roller bottles, DURAN®
Fournisseur: Carl Roth
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ROTHT967.1EA 0 EUR
ROTHT967.1
Cell culture roller bottles, DURAN®
Flacons Flacons Roller
DURAN®, borosilicate glass with DIN thread (GL 45)
  • Very good temperature resistance
  • Very good chemical resistance
Supplied with blue PP screw cap and pouring ring.