Imprimer
Cartridge filters, Optimizer® D300 and D600
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CWAG031S2 CWAG061S2 CWAX031S2 CWAX061S2 CWAZ061S2 CWAT061S2 CWMX031S2 CWAV061S2
ENTECWAG031S2EA 0 EUR
ENTECWAG031S2 ENTECWAG061S2 ENTECWAX031S2 ENTECWAX061S2 ENTECWAZ061S2 ENTECWAT061S2 ENTECWMX031S2 ENTECWAV061S2
Cartridge filters, Optimizer® D300 and D600
Filtres Cartouches filtrantes
The point-of-use photochemical filters with superior contaminant retention and low hold-up volume performance in sub-10 to 180 nm technology nodes. Both Optimizer® D300 and D600 filters are geared to small-volume filtration of photoresists, resist dielectrics, and solvents. It requires no pre-wetting, so the filters function immediately after installation. Without the need for prewetting, they reduce the likelihood that microbubbles will form and avoid the problems associated with incomplete wetting and the mixing of incompatible chemicals.
  • Suitable for semiconductor industry, photolithography, chemical manufacturing and DI water
Membrane: hydrophobic UPE; Supports, housing: HDPE
Maximum operating temperature: 60 °C
Maximum forward/reverse differential pressure: 2,4 bar, 35 psid at 25 °C
Maximum operating pressure: 3,4 bar, 50 psid at 25 °C