Imprimer
HPLC columns, Hibar®, LiChrospher® 100 NH₂
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.00423.7076 1.00424.7076 1.00424.7077 1.50009.7076 1.50009.7077 1.50012.7076 1.50013.7076 1.50013.7077 1.50182.7076 1.50182.7077 1.50183.7076 1.50183.7077
1.00423.7076EA 0 EUR
1.00423.7076 1.00424.7076 1.00424.7077 1.50009.7076 1.50009.7077 1.50012.7076 1.50013.7076 1.50013.7077 1.50182.7076 1.50182.7077 1.50183.7076 1.50183.7077
HPLC columns, Hibar®, LiChrospher® 100 NH₂
Colonnes pour chromatographie
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.