Imprimer
Cartridge filters, Optimizer® DEV and DI
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CBUG40KL1 CBUG16KL1 CBUG40EL1 CBUV40EL1 CBUZ16KL1 CBUZ40KL1 CHUV16KL1 CHUV40EL1 CHUV40KL1 CHUV40KM1 CHUZ40KL1
ENTECBUG40KL1EA 0 EUR
ENTECBUG40KL1 ENTECBUG16KL1 ENTECBUG40EL1 ENTECBUV40EL1 ENTECBUZ16KL1 ENTECBUZ40KL1 ENTECHUV16KL1 ENTECHUV40EL1 ENTECHUV40KL1 ENTECHUV40KM1 ENTECHUZ40KL1
Cartridge filters, Optimizer® DEV and DI
Filtres Cartouches filtrantes
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, end caps, cage and core: HDPE.
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,7 bar at 25 °C
Maximum reverse differential pressure: 2,7 bar at 25 °C