Imprimer
Point of use photochemical filters, Optimizer®
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABUG0SKM1 ABUZ0LKM1 ABUZ0SKM1 AHUV0LKM1 AHUV0SKM1 AHUYMLKM1 AHUZ0LKM1 AHUZ0SKM1 AWAV0SKM1 AWAXMSKM1
ENTEABUG0SKM1EA 0 EUR
ENTEABUG0SKM1 ENTEABUZ0LKM1 ENTEABUZ0SKM1 ENTEAHUV0LKM1 ENTEAHUV0SKM1 ENTEAHUYMLKM1 ENTEAHUZ0LKM1 ENTEAHUZ0SKM1 ENTEAWAV0SKM1 ENTEAWAXMSKM1
Point of use photochemical filters, Optimizer®
Filtres Cartouches filtrantes
Liquid photochemical filters designed for developer, DI water and solvent applications in POU photolithography processes.
  • Advanced filtration media: optimises filtration and particle retention capabilities
  • UPE membrane spontaneously wets and stays wet in solvents
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UPE); Supports, core and sleeve: HDPE; Housing: PP
Maximum operating temperature: 40 °C
Maximum inlet pressure: 3,9 bar at 25 °C
Maximum forward/reverse pressure: 2,8 bar at 25 °C