Imprimer
Magnetic stirring bars, cylindrical, PTFE, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F371110001 F371110002
BELAF371110001EA 0 EUR
BELAF371110001 BELAF371110002
Magnetic stirring bars, cylindrical, PTFE, Spinbar®, SP Bel-Art
Agitateurs Barreaux magnétiques
Stirring bars ensure centering and smooth running characteristics via a small removable pivot ring in the center. The pivot ring minimises the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers.