Imprimer
Oil mist eliminators
Fournisseur: ALCATEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
066849 104200 104199 068785
ALCA066849EA 0 EUR
ALCA066849 ALCA104200 ALCA104199 ALCA068785
Oil mist eliminators
Filtres Filtres à gaz
Standard series:
These models can retain oil droplets smaller than 1 μm in diameter. They feature an internal over pressure valve which prevents exceeding the maximum permissible exhaust pressure.
Chemical series:
These models are suitable for mechanical pumps. Oil that condenses does not recycle back to the pump but remains in the mist eliminator body. They feature a sight glass, which enables the operator to monitor the quantity of oil retained.