Imprimer
Total organic carbon (TOC) vials
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40C-TOC/LL 40C-TOC/DB/LL 40C-TOC 40C-TOC/DB
EPSC40C-TOC/LLEA 824 EUR
EPSC40C-TOC/LL EPSC40C-TOC/DB/LL EPSC40C-TOC EPSC40C-TOC/DB
Total organic carbon (TOC) vials
Fioles
Total Organic Carbon (TOC), certified vials
  • USP Method 643 Testing
  • Off-line and grab sampling of high purity water
  • Low background is perfect for preparation and storage of standards
  • Cleaning validation is simplified and costs are reduced by using the TOC method of testing