Imprimer
Tool kit
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PERCN9301328EA 1210 EUR
PERCN9301328
Tool kit
Sets d'outils
Kit includes the following tools: 6 × openend wrench set, 6 × screwdriver set, adjustable wrench (6" ), chain nose pliers (narrow), wire cutters, wire strippers, slip-joint pliers (6"), long nose No. 5 stainless steel tweezers (4 ³/₈"), 6 × needle file set, 11 × xAllen key set (imperial sizes), 9 × Allen key set (metric sizes). Shipped in a plastic tool box for convenient storage.