Imprimer
Multi-tube racks, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F185132040 F185131640 F185130072
BELAF185132040EA 101 EUR
BELAF185132040 BELAF185131640 BELAF185130072
Multi-tube racks, SP Bel-Art
Portoirs Tube Racks
PP
  • Chemical or corrosion resistant
  • Very good temperature resistance
  • End plates have hand holes for safe transport
Ideal for serum vials, RIA vials, T3/T4 reaction vials. It can be used in water baths for elevated temperature incubations.