Imprimer
Measuring cylinders, single scale graduated, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F284576000 F284580000 F284550000
BELAF284576000EA 51.2 EUR
BELAF284576000 BELAF284580000 BELAF284550000
Measuring cylinders, single scale graduated, SP Bel-Art
Éprouvettes Éprouvettes graduées
PP, tall form.
  • Easy-to-read, no meniscus, molded-in graduations
  • Excellent chemical resistance
  • Polygon base, roll proof