Imprimer
Glassware draining racks, electric, benchtop, Lab-Aire® II, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F188192304 F188192301
BELAF188192304EA 5830 EUR
BELAF188192304 BELAF188192301
Glassware draining racks, electric, benchtop, Lab-Aire® II, SP Bel-Art
Portoirs Égouttoirs
ABS plastic.
  • Heat selector knob with three temperature settings: high, medium and low
  • Independent thermo-fuse protects unit from overheating
Electric benchtop dryer offer a compact footprint and accelerate drying of labware with warm, circulating air. Electric drying efficiency is enhanced with a replaceable polyurethane foam filter that keeps warmed air dust free.