Imprimer
Culture media in bags, ReadyBags™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BM0104T BM0204T BM1104T BM9901A
OXOIBM0104TEA 293 EUR
OXOIBM0104T OXOIBM0204T OXOIBM1104T OXOIBM9901A
Culture media in bags, ReadyBags™
Milieux pour microbiologie Milieux prêts à l'emploi de microbiologie
ReadyBags™ are designed for use in food testing by laboratories with large numbers of samples requiring large volumes of broth media or diluent. Each ReadyBag™ contains 3 liters and is intended for use with an automatic dispenser. The ReadyBag™ is connected to tubing for standard laboratory equipment, such as gravimetric diluters or peristaltic pumps, by means of a connector. Connectors are available in two formats: autoclavable, re-usable stainless steel connectors or sterile, plastic, single-use connectors.
  • For large numbers of samples
  • Available in 3 L
  • Once empty, the bag can be disposed as normal laboratory waste