Imprimer
Bulb pipettes (one mark), BLAUBRAND®, Class AS, LOT certificate (one batch)
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.29704 2.29707 2.29712 2.29713 2.29705 2.29706 2.29708 2.29709 2.29710 2.29711 2.29717 2.29701 2.29702 2.29714 2.29703 2.29716 2.29715 2.29719 2.29718
BRND2.29704EA 57.9 EUR
BRND2.29704 612-1063 612-1064 BRND2.29713 BRND2.29705 BRND2.29706 BRND2.29708 BRND2.29709 BRND2.29710 BRND2.29711 BRND2.29717 BRND2.29701 BRND2.29702 BRND2.29714 BRND2.29703 BRND2.29716 BRND2.29715 BRND2.29719 BRND2.29718
Bulb pipettes (one mark), BLAUBRAND®, Class AS, LOT certificate (one batch)
Pipettes Pipettes jaugées
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
  • All supplied pipettes come from one batch for statistical testing equipment monitoring
  • DE-M marking, with batch certificate and LOT certificate
Certifications: DIN EN ISO 648; DE-M marking.