Imprimer
Tissue grinders, DUALL®, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP885480-0020 SP885480-0022 SP885480-0023
SCERSP885480-0020EA 267 EUR
SCERSP885480-0020 SCERSP885480-0022 SCERSP885480-0023
Tissue grinders, DUALL®, KIMBLE®
Homogénéisateurs Broyeurs pour tissus
These tissue grinders (borosilicate glass, PTFE pestle with glass tube) combine both conical and cylindrical surfaces to reduce tissue and also produces a uniform homogenate. Construction is strong enough to allow the pestle to be motor driven and also the grinding efficiency is greatly improved while using this tube.
  • Replacement components are available and completely interchangeable
  • Used for soft tissue such as brain and liver
Clearance between pestles and tubes is 0,004 to 0,006"