Imprimer
NMR sample tubes, 5 mm, disposable, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP897193-0000 SP897193-0008 SP897193-7100 SP897193-8100
SCERSP897193-0000EA 307 EUR
SCERSP897193-0000 SCERSP897193-0008 SCERSP897193-7100 SCERSP897193-8100
NMR sample tubes, 5 mm, disposable, KIMBLE®
Tubes Tubes RMN
Borosilicate glass.
External diameter: 5 mm
Wall thickness: 0,381 mm
Informations de livraison: Including PE cap which are not attached.