Imprimer
Chromatographic columns with frit, PTFE stopcock, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP420550-0256 SP420550-0430
SCERSP420550-0256EA 960 EUR
SCERSP420550-0256 SCERSP420550-0430
Chromatographic columns with frit, PTFE stopcock, KIMBLE®
Colonnes pour chromatographie
These borosilicate glass gravity columns are manufactured from medium wall tubing with PTFE stopcock. These columns have a coarse porosity glass frit (40 - 60 μm pore size) at the lower end to act as a support for column packing material. Also these columns can be used alone or in combination with reservoirs and flow control adapters.
Informations de commande: Custom sizes and plastic safety coating are available upon request.