Imprimer
Distilling bridges, Claisen
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05057314 05057211 05057311 05057414
LENZ05057314EA 208 EUR
LENZ05057314 LENZ05057211 LENZ05057311 LENZ05057414
Distilling bridges, Claisen
Distillation Components Distillation Bridges
DURAN®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling link with Liebig condenser and Claisen head. With two sockets NS 14/23 for distillation thermometer of 55 mm buli-in length. With two thread connections, GL 14 (water) and GL 18 (vacuum).