Imprimer
Wide opening snap cap vial closures, Target® Snap-It™
Fournisseur: THERMO NATIONAL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C4011-51G C4011-53 C4011-53Y C4011-52B C4011-52G C4011-53B C4011-52R
NSCAC4011-51GEA 44.6 EUR
NSCAC4011-51G NSCAC4011-53 NSCAC4011-53Y NSCAC4011-52B NSCAC4011-52G NSCAC4011-53B NSCAC4011-52R
Wide opening snap cap vial closures, Target® Snap-It™
Capuchons Bouchons pour flacons de chromatographie
These chemically inert PE or PP caps are suitable for most chromatography applications and are an economical choice for routine HPLC, GC and IC applications.
  • Low sealing property and zero resealing capacity
  • Easy to apply and easy to remove and fully compatible with all Snap-It™ vials
  • Enlarged open area allows for needle penetration across the entire vial opening