Imprimer
Handheld digital microscopes, Dino-Lite High Magnification
Fournisseur: Dino-Lite / IDCP B.V.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AM4013MZT4 AM4113ZT4 AM4013MT5 AM4113T5 AM4515T5 AM4515T8 AM4515ZT4 AM7515MT4A AM7515MT8A AM73515MT8A
IDCPAM4013MZT4EA 695 EUR
IDCPAM4013MZT4 IDCPAM4113ZT4 IDCPAM4013MT5 IDCPAM4113T5 IDCPAM4515T5 IDCPAM4515T8 630-2750 IDCPAM7515MT4A IDCPAM7515MT8A IDCPAM73515MT8A
Handheld digital microscopes, Dino-Lite High Magnification
Microscopes
Dino-Lite models within the High Magnification series exceed a magnification of 200 times and offer magnifications of 400×, 500× or even 900 times. Magnification range: 400 - 900×.
  • High speed USB 2.0 connection
  • Metal housing or composite housing
  • Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Coaxial models: The coaxial illumination technique reveals details that are very difficult to see under normal light, for example when inspecting wafer plates, microchips or other microelectronics. In combination with the Flexible LED Control (FLC), these models are capable of switching between (or mixing) brightfield and coaxial illumination.
Dino-Lite High Magnification models are great inspection tools for biomedical and scientific research, material analysis, electronics inspection, or any similar application that requires high magnification, versatility and mobility.
Informations sur les accessoires: Please note: When focusing at such high magnification it is recommended to use a high-precision stand (e.g. Dino-Lite RK-10A).
Informations de livraison: Supplied with carry pouch, Software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), user manual, Quick guide and calibration target.