Imprimer
Analytical chromatography accessories for chiral columns
Fournisseur: CHIRAL TECHNOLOGIES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
00025 00024 00011
DAIC00025EA 135 EUR
DAIC00025 DAIC00024 DAIC00011
Analytical chromatography accessories for chiral columns
Colonnes pour chromatographie
The HPLC analytical guard cartridges need to be mounted in a cartridge holder to be connected to the analytical column. For maximum lifetime, analytical columns should be used in series with an appropriate guard cartridge or column. Both formats are available with the packing material to match the protected column.
Column jackets allow operating temperatures of analytical columns to be controlled with a circulating water bath.