Imprimer
Reagents for determination of silica
Marques : Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
199532 27032 199553 2254249 199549
HACH199532EA 40.5 EUR
HACH199532 HACH27032 HACH199553 HACH2254249 HACH199549
Reagents for determination of silica
Kits de tests environnementaux
These reagents used for low and ultra-low range silica determination by the Heteropoly Blue method.