Imprimer
Heat-shrink bands for PETG media bottles, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312160-0384 312160-0240 312160-0530 312160-0200
NALG312160-0384EA 156 EUR
NALG312160-0384 NALG312160-0240 NALG312160-0530 NALG312160-0200
Heat-shrink bands for PETG media bottles, Nalgene®
Scellés Scellés de sécurité
These heat-shrink bands can be used to ensure the integrity of bottle contents. Simply apply the recommended application torque for Thermo Scientific Nalgene® closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.
  • Gamma stable bands include a perforated "tear strip" feature for easy removal
Cdt: Bands are packed 1000 per case (two zipper bags of 500 each).