Imprimer
Buffer solutions in sachets, 25 °C, AVS TITRINORM®
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
85042.001EA 116 EUR
85042.001 85041.001 85043.001 85044.001 85045.001 85046.001 85041.002 85042.002 85043.002 85044.002 85045.002 85046.002
Buffer solutions in sachets, 25 °C, AVS TITRINORM®
Buffers pH Calibration Buffers
For calibration of pH meters.
  • Practical colour coded 30 ml sachets - one use per sachet - no risk of contamination
  • Ideal for on-site analyses
  • Accuracy: pH is ±0,02 at 25 °
  • Traceable to SRM from NIST
Cdt: Two versions available: 30× 30 ml in a standard box and 30× 20 ml in a user-friendly dispenser box