Imprimer
Kjeldahl tablets
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.10958.0250 1.10958.1000 1.15348.0250 1.15348.1000 1.17958.0250 1.16469.0250 1.16469.1000 1.18348.0250 1.18469.0250
1.10958.0250EA 380 EUR
1.10958.0250 1.10958.1000 1.15348.0250 1.15348.1000 1.17958.0250 1.16469.0250 1.16469.1000 1.18348.0250 1.18469.0250
Kjeldahl tablets
Digestion Systems Kjeldahl Tablets and Reagents
In addition to the traditional 5 g Kjeldahl tablets, which are added as catalysts to the digestion mixture, Merck Millipore can offer the 3,5 g tablet. This gives the flexibility needed to find the optimal setting for the catalyst/sulphuric acid ratio.
  • The 3,5 g tablets are suitable for microdigestion
  • Easy to use PE packaging concept