Imprimer
Stoppers, hollow, LDPE
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1448001 1448000
HACH1448001EA 23.3 EUR
HACH1448001 HACH1448000
Stoppers, hollow, LDPE
Bouchons Bouchons en plastique
Ideally can be used as a small beaker, sample cup for pH meters, flame photometers and automatic analysers.
  • LDPE
  • Interchangeable with rubber stoppers
  • Safer and economical
  • Fits snugly