Imprimer
Reagent powder pillows for determination of molybdenum, MolyVer®
Marques : Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1414669 1414869 1417869 2604399 2604499
HACH1414669EA 86.3 EUR
HACH1414669 HACH1414869 HACH1417869 HACH2604399 HACH2604499
Reagent powder pillows for determination of molybdenum, MolyVer®
Kits de tests environnementaux
For determination of high range molybdenum by the mercaptoacetic acid method.