Visual comparator test kits, copper
Marques : HachTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Simple, economical visual test kit method for copper.
Informations de livraison: Kit includes reagents, colour disc, comparator box, two viewing tubes, instructions and carrying case.