Imprimer
Film applicators, multi-functional
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VF2174 VF2175 VF2173 VF2172 VF2170 VF2169 VF2168 VF2167
SIMEVF2174EA 740 EUR
SIMEVF2174 SIMEVF2175 SIMEVF2173 SIMEVF2172 SIMEVF2170 SIMEVF2169 SIMEVF2168 SIMEVF2167
Film applicators, multi-functional
Film Applicators Paint/Ink Film Applicators
Multi-functional film applicator with 4 application sides for applying paint films of 4 different predefined thicknesses. The applicators are available in film widths of 60, 80 mm and one side of the applicator is supplied with a guidance support for straight application. Made from high grade stainless steel, the multi-functional film applicator will not be affected by acid or base elements.