Imprimer
Retort clamps
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5464 5480 5481 5482
BOCH5464EA 27.7 EUR
BOCH5464 BOCH5480 BOCH5481 BOCH5482
Retort clamps
Pinces Pinces à usage général
Steel, zinc plated.
  • Round jaws with cork lining to protect clamped items
  • Without bosshead for fixing with stand
  • With wing screws for rapid easy clamping
DIN 12894
Certifications: DIN 12894