Imprimer
Disinfectant and precleaner, ANIOS'CLEAN EXCEL D
Fournisseur: Laboratoires Anios
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
24160366C 2416095 2416097
DMDA24160366CEA 690 EUR
DMDA24160366C DMDA2416095 DMDA2416097
Disinfectant and precleaner, ANIOS'CLEAN EXCEL D
Produits nettoyants Désinfectants
Designed for cleaning and pre-disinfection of surgical instrumentation, medical devices, heat-sensitive instruments and endoscopy equipment.
  • High level of detergency shown
  • Active on BMR and BHR
  • Effective even in hard water
  • Non corrosive, compatible with a wide range of materials
Certifications: Bactericidal: EN 1040, EN 13727; Fungicidal: EN 1275, EN 13624, EN 14562, EN 1650; Mycobactericidal: EN 14348, EN 14563; Virucidal: HIV-1, PRV, BVDV, HSV-1, Vacciniavirus.