Imprimer
Frits for suction filters, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F766-08 F766-48
BOHLF766-08EA 126 EUR
BOHLF766-08 BOHLF766-48
Frits for suction filters, BOLA
Filtres Filtres de précolonnes
These frits are made from PTFE.
  • Temperature resistant from −200 to +250 °C
  • Chemically resistant
  • Conform to FDA