Imprimer
Universal containers with slip-over lid, UNICON
Fournisseur: schuett-biotec GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3526113 3526173 3526183 3526273
489-1870EA 105 EUR
489-1870 489-1871 489-1872 489-1873
Universal containers with slip-over lid, UNICON
Récipients de collecte d'échantillon Conteneurs à usage universel
These round universal containers are made of pure aluminium.
  • Lightweight
  • Sturdy
  • Easy to clean
They have a slip-over lid, which is ideal for storing and sterilisation of pipette tips and all other small parts made of plastic or glasses.