Imprimer
Bottle carrier, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F169590000 F169570000 F169580000 F169600000
BELAF169590000EA 362 EUR
BELAF169590000 BELAF169570000 BELAF169580000 BELAF169600000
Bottle carrier, SP Bel-Art
Portoirs Portoirs pour flacons
HDPE.
  • Epoxy coating
  • Heavy-gauge steel wire bail handle
  • Side ribs and bottom suspend the filled bottle
These bottle carriers are suitable for transport and storage of reagent bottles. The cone-shaped opening allows a delivery tube to be inserted, so liquid does not have to be poured out. The shortened lid allows the bottle to protrude for safer pouring.
Colour: White.