Imprimer
Single wafer tray, H 20 series, 200 mm
Fournisseur: Entegris
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ENTEH207000011415EA 0 EUR
ENTEH207000011415
Single wafer tray, H 20 series, 200 mm
Matériel de manipulation de wafers Plateaux pour semi-conducteurs
Wafer handling trays to safely transport and protect silicon wafers.
  • For internal handling of single wafers
  • Conical pocket holds one wafer facedown contacting only the wafer’s edges
  • Tweezer slots on tray bottom ease wafer removal
  • Stackable for efficient use of storage space, either tray on tray or cover on tray