Imprimer
Single wafer tray, H 20 series, 150 mm
Fournisseur: Entegris
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ENTEH205000011415EA 0 EUR
ENTEH205000011415
Single wafer tray, H 20 series, 150 mm
Matériel de manipulation de wafers Plateaux pour semi-conducteurs
To support the semiconductor industry's requirements for advanced 150 mm wafer handling, Entegris offers a variety of wafer shipping and handling solutions to safely transport silicon wafers in today's 150 mm fabs.
  • For internal handling and storage applications
  • Holds one wafer face down in a conical pocket contacting only the wafer’s edges
  • Tweezer slots conveniently located for easy wafer handling
  • Stackable tray on tray for efficient use of storage space