Imprimer
Proteomic protein controls
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PSC-01 PSC-02 PSC-03 PSC-04 PSC-05
PSC-01EA 305 EUR
PSC-01 PSC-02 PSC-03 PSC-04 PSC-05
Proteomic protein controls
Protéines purification Réactifs de purification des protéines
Protein preparations specifically prepared for standardising electrophoresis methods and protocols. Substantially free from non-protein agents such as nucleic acids, detergents, salts, lipids and other common laboratory agents, the protein preparation has low conductivity (<50 μs). Supplied as dry protein pellets, simply re-hydrate and use. Proteomic protein sample controls are available for animal cells, E.coli, yeast, plant or as a control set that contains one 2 mg vial of each.