Imprimer
Safety dispensing canisters
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16780 16781
MSGA16780EA 831 EUR
MSGA16780 MSGA16781
Safety dispensing canisters
Safety Cans
Canister, made of stainless steel, with tap and separate ventilation.
  • Manual pressure
  • Vacuum balancing
  • Self-closing tap