Imprimer
Bag closures with scoop/spoon, WeLoc® Scoop
Fournisseur: WELOC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S421100150404 S421500550404 S422201100404 S42320250040
312-0115EA 50.1 EUR
312-0115 312-0116 312-0117 312-0118
Bag closures with scoop/spoon, WeLoc® Scoop
Clamps General Purpose Clamps
These sealing clips feature an integrated measuring scoop/spoon. They are convenient for many types of applications in the domestic, medical and industrial field, where both a seal and a measure is required. The clips are made from high quality Polyamide 6.6 material from DuPont® and therefore perfect for use in the demanding pharmaceutical and laboratory environment.
  • Re-closing of partly used bags of ingredients will reduce the risk of cross-contamination
  • Reduce the risk of leakage or accidents
  • Made from strong and durable material
  • Manufactured in Sweden under strict hygiene conditions
  • Clips can be sterilised via autoclave, EtO or Gamma systems before being re-used