Imprimer
Single wafer shippers for wafer size 76,2 mm (3") or 90 mm
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H22-301-0615 H22-30-0615_U H22-302-0815 H22-302A-0615 H22-302-0615 H22-321-0615
216-1747EA 0 EUR
216-1747 ENTEH22-30-0615_U ENTEH22-302-0815 216-2364 216-1748 ENTEH22-321-0615
Single wafer shippers for wafer size 76,2 mm (3") or 90 mm
Wafer Handling Supplies Wafer Shippers
Conical pocket holds one wafer face down contacting only the wafer edges.
  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
  • Large area on cover provides space for labelling
Attention: Articles may not be available in all countries. Please contact your local Avantor office.