Imprimer
Couvercles de plateaux pour puces 4", série H44
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H44-02LR-66C02 H44-02A-5413 H44-02B-66C02 H44-02B-1216 H44-02C-62C02 H44-02C-66C02 H44-02-1201 H44-02-1216 H44-02-1415_U H4402-15-74D10 H44-02-74D10 H44-02A-62C02 H44-02A-66C02 H44-02B-62C02 H4402RM66C02
ENTEH44-02LR-66C02EA 0 EUR
ENTEH44-02LR-66C02 ENTEH44-02A-5413 ENTEH44-02B-66C02 ENTEH44-02B-1216 ENTEH44-02C-62C02 ENTEH44-02C-66C02 ENTEH44-02-1201 ENTEH44-02-1216 ENTEH44-02-1415_U ENTEH4402-15-74D10 ENTEH44-02-74D10 ENTEH44-02A-62C02 ENTEH44-02A-66C02 ENTEH44-02B-62C02 ENTEH4402RM66C02
Couvercles de plateaux pour puces 4", série H44
Couvercles Lids for Laboratory Vessels/Containers
Tray covers in various standard materials are shown below.
  • STAT-PRO® 150 covers and trays are not recommended for use with devices less than .012"
  • STAT-PRO® 150 material covers are recommended for use only with STAT-PRO® 150 material trays
Articles may not be available in all countries. Please contact your local Avantor office.