Imprimer
Sacs, Hazard BinLiner™
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BL2-25REIRBB BL2-32YEIRBB BL4-32YEIRBB BH2-24YEIRBB
129-1088EA 324 EUR
129-1088 129-1597 129-1589 129-1653
Sacs, Hazard BinLiner™
Sachets Sacs à déchets
These low density polyethylene Hazard BinLiner™ bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
  • Low density polyethylene
  • Hazardous material bags are labelled for waste segregation in the U.S.
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Can be used with BLSSH series BinLiner™ Stands
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
The Biohazard liners are designed for containment and disposal of biomedical waste and the Hazardous Material liners are designed for segregation and disposal of hazardous waste. Biohazard bags are clearly marked with a Biohazard symbol in black ink.
Certifications: Certificate of Irradiation and Certificate of Conformance shipped with each order. Validation paperwork available upon request. Minimum irradiation dose: 25 kGy.