Imprimer
Tubing connectors
Fournisseur: CADENCE SCIENCE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6169IND 6184
POPR6169INDEA 34.3 EUR
POPR6169IND POPR6184
Tubing connectors
Raccords et connecteurs
These tubing connectors are used in hospitals, laboratories, and biomedical instruments. The metal connectors are precision machined and are made from either nickel plated brass or 316 stainless steel.
Standard luer tapers meets ISO 594 specifications include Female Luer Lock (FLL), Micro-Mate® Female Luer Lock (MM FLL), Male Luer Lock (MLL) and Male Luer (ML).